Assay Details
| Gene Symbol: TMEM106B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283071.1 | NP_001270000.1 | 8 - 9 | 933 | 120 | |
| XM_005550096.2 | XP_005550153.1 | 7 - 8 | 909 | 120 | ||
| XM_005550100.2 | XP_005550157.1 | 7 - 8 | 940 | 120 | ||
| XM_015447278.1 | XP_015302764.1 | 7 - 8 | 1365 | 120 | ||
| GenBank mRNA | AB172352.1 | - | 8 - 9 | 933 | 120 | |
More Information
Set Membership: |
|

TMEM106B
Genomic Map