Assay Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575448.2 | XP_005575505.1 | 4 - 5 | 1203 | 76 | |
| XM_005575450.2 | XP_005575507.1 | 4 - 5 | 972 | 76 | ||
More Information
Set Membership: |
|

TRABD2A
Genomic Map