Assay Details
| Gene Symbol: CADM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283689.1 | NP_001270618.1 | 3 - 4 | 692 | 82 | |
| GenBank mRNA | AB172671.1 | - | 3 - 4 | 692 | 82 | |
More Information
Set Membership: |
|

CADM3
Genomic Map