Assay Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283986.1 | NP_001270915.1 | 5 - 6 | 536 | 84 | |
| GenBank mRNA | AB171940.1 | - | 5 - 6 | 536 | 84 | |
More Information
Set Membership: |
|

ZNF330
Genomic Map