Assay Details
| Gene Symbol: EFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560278.2 | XP_005560335.1 | 17 - 18 | 2205 | 95 | |
| XM_015453174.1 | XP_015308660.1 | 18 - 19 | 2259 | 95 | ||
| XM_015453175.1 | XP_015308661.1 | 17 - 18 | 2121 | 95 | ||
More Information
Set Membership: |
|

EFL1
Genomic Map