Assay Details
| Gene Symbol: FSHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284639.1 | NP_001271568.1 | 2 - 3 | 218 | 91 | |
| XM_005575919.2 | XP_005575976.1 | 2 - 3 | 328 | 91 | ||
| GenBank mRNA | X74454.2 | - | 218 | 91 | ||
More Information
Set Membership: |
|

FSHR
Genomic Map