Assay Details
| Gene Symbol: SLC22A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284124.1 | NP_001271053.1 | 7 - 8 | 1284 | 115 | |
| XM_005577402.2 | XP_005577459.1 | 6 - 7 | 1562 | 115 | ||
| XM_005577403.2 | XP_005577460.1 | 6 - 7 | 1538 | 115 | ||
| XM_005577410.2 | XP_005577467.1 | 6 - 7 | 1027 | 115 | ||
| XM_015434191.1 | XP_015289677.1 | 5 - 6 | 1643 | 115 | ||
| XM_015434193.1 | XP_015289679.1 | 6 - 7 | 1763 | 115 | ||
| XM_015434194.1 | XP_015289680.1 | 5 - 6 | 1677 | 115 | ||
| GenBank mRNA | AB738914.1 | - | 7 - 8 | 1284 | 115 | |
More Information
Set Membership: |
|

SLC22A12
Genomic Map