Assay Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588893.2 | XP_005588950.1 | 8 - 9 | 1018 | 62 | |
| XM_005588894.2 | XP_005588951.1 | 8 - 9 | 881 | 62 | ||
| XM_015441598.1 | XP_015297084.1 | 9 - 10 | 941 | 62 | ||
More Information
Set Membership: |
|

LIN37
Genomic Map