Assay Details
Gene Symbol: C1QTNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556669.2 | XP_005556726.1 | 2 - 3 | 1814 | 90 | |
XM_005556671.2 | XP_005556728.1 | 2 - 3 | 1586 | 90 | ||
XM_005556672.2 | XP_005556729.1 | 2 - 3 | 217 | 90 | ||
XM_005556673.2 | XP_005556730.1 | 2 - 3 | 262 | 90 | ||
XM_015451257.1 | XP_015306743.1 | 2 - 3 | 1774 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
