Assay Details
| Gene Symbol: SLC41A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284000.1 | NP_001270929.1 | 6 - 7 | 1177 | 65 | |
| XM_015430636.1 | XP_015286122.1 | 5 - 6 | 1191 | 65 | ||
| XM_015430637.1 | XP_015286123.1 | 5 - 6 | 1351 | 65 | ||
| XM_015430638.1 | XP_015286124.1 | 6 - 7 | 1271 | 65 | ||
| XM_015430639.1 | XP_015286125.1 | 6 - 7 | 1466 | 65 | ||
| XM_015430640.1 | XP_015286126.1 | 5 - 6 | 1133 | 65 | ||
| XM_015430641.1 | XP_015286127.1 | 5 - 6 | 1211 | 65 | ||
| XM_015430642.1 | XP_015286128.1 | 4 - 5 | 1083 | 65 | ||
| GenBank mRNA | AB179433.1 | - | 6 - 7 | 1177 | 65 | |
More Information
Set Membership: |
|

SLC41A2
Genomic Map