Assay Details
| Gene Symbol: TMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585115.2 | XP_005585172.1 | 8 - 9 | 1805 | 65 | |
| XM_005585116.2 | XP_005585173.1 | 8 - 9 | 1765 | 65 | ||
| XM_015438901.1 | XP_015294387.1 | 8 - 9 | 1774 | 65 | ||
| XM_015438902.1 | XP_015294388.1 | 8 - 9 | 1776 | 65 | ||
| XM_015438903.1 | XP_015294389.1 | 5 - 6 | 585 | 65 | ||
More Information
Set Membership: |
|

TMC8
Genomic Map