Assay Details
| Gene Symbol: POC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285163.1 | NP_001272092.1 | 10 - 11 | 1613 | 79 | |
| XM_005557193.2 | XP_005557250.1 | 11 - 12 | 1782 | 79 | ||
| XM_005557194.2 | XP_005557251.1 | 11 - 12 | 1894 | 79 | ||
| GenBank mRNA | AB168488.1 | - | 10 - 11 | 1613 | 79 | |
More Information
Set Membership: |
|

POC5
Genomic Map