Assay Details
| Gene Symbol: CLDN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447193.1 | XP_015302679.1 | 3 - 4 | 2232 | 95 | |
| XM_015447194.1 | XP_015302680.1 | 3 - 4 | 2192 | 95 | ||
More Information
Set Membership: |
|

CLDN15
Genomic Map