Assay Details
| Gene Symbol: MMP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568817.2 | XP_005568874.2 | 7 - 8 | 1624 | 64 | |
| XM_005568818.2 | XP_005568875.1 | 6 - 7 | 1643 | 64 | ||
| XM_005568819.2 | XP_005568876.1 | 5 - 6 | 1481 | 64 | ||
More Information
Set Membership: |
|

MMP24
Genomic Map