Assay Details
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552499.2 | XP_005552556.1 | 5 - 6 | 1341 | 88 | |
| XM_005552500.1 | XP_005552557.1 | 5 - 6 | 1407 | 88 | ||
| XM_005552501.2 | XP_005552558.1 | 5 - 6 | 1551 | 88 | ||
| GenBank mRNA | AB063014.1 | - | 5 - 6 | 1363 | 88 | |
More Information
Set Membership: |
|

LCA5
Genomic Map