Assay Details
Gene Symbol: ICMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544934.2 | XP_005544991.1 | 1 - 2 | 264 | 67 | |
XM_015443375.1 | XP_015298861.1 | 1 - 2 | 262 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
