Assay Details
| Gene Symbol: MYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284979.1 | NP_001271908.1 | 1 - 2 | 224 | 61 | |
| GenBank mRNA | AB171006.1 | - | 1 - 2 | 224 | 61 | |
More Information
Set Membership: |
|

MYG1
Genomic Map