Assay Details
Gene Symbol: RAB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283960.1 | NP_001270889.1 | 4 - 5 | 493 | 78 | |
GenBank mRNA | AB169319.1 | - | 4 - 5 | 490 | 78 | |
AB169768.1 | - | 4 - 5 | 493 | 78 | ||
AB170418.1 | - | 4 - 5 | 331 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
