Assay Details
| Gene Symbol: NGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542226.2 | XP_005542283.1 | 2 - 3 | 157 | 78 | |
| XM_015430630.1 | XP_015286116.1 | 1 - 2 | 10443 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005542226.2 transcripts.
Set Membership: |
|

NGF
Genomic Map