Assay Details
| Gene Symbol: HTR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574572.2 | XP_005574629.1 | 2 - 3 | 570 | 71 | |
| XM_015432943.1 | XP_015288429.1 | 1 - 2 | 1654 | 71 | ||
More Information
Set Membership: |
|

HTR2B
Genomic Map