Assay Details
Gene Symbol: VCPKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005561215.2 | XP_005561272.1 | 2 - 3 | 881 | 67 | |
XM_005561216.2 | XP_005561273.1 | 2 - 3 | 881 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
