Assay Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571779.2 | XP_005571836.1 | 3 - 4 | 1464 | 103 | |
| XM_005571780.2 | XP_005571837.1 | 2 - 3 | 1566 | 103 | ||
| XM_005571781.2 | XP_005571838.1 | 2 - 3 | 1566 | 103 | ||
| XM_005571782.2 | XP_005571839.1 | 2 - 3 | 1566 | 103 | ||
| XM_015431359.1 | XP_015286845.1 | 2 - 3 | 1566 | 103 | ||
More Information
Set Membership: |
|

KBTBD12
Genomic Map