Assay Details
Gene Symbol: NOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283542.1 | NP_001270471.1 | 7 - 8 | 840 | 69 | |
GenBank mRNA | AB169707.1 | - | 7 - 8 | 840 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
