Assay Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015451643.1 | XP_015307129.1 | 2 | 2309 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD34B| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015451643.1 | XP_015307129.1 | 2 | 2309 | 133 | |
Set Membership: |
|