Assay Details
| Gene Symbol: OPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589596.2 | XP_005589653.1 | 1 - 2 | 304 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OPA3| Gene Symbol: OPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589596.2 | XP_005589653.1 | 1 - 2 | 304 | 84 | |
Set Membership: |
|