Assay Details
| Gene Symbol: CYP26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565962.2 | XP_005566019.1 | 1 | 216 | 103 | |
| XM_015456398.1 | XP_015311884.1 | 1 | 216 | 103 | ||
More Information
Set Membership: |
|

CYP26A1
Genomic Map