Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588909.2 | XP_005588966.1 | 3 - 4 | 176 | 74 | |
| XM_005588910.2 | XP_005588967.1 | 3 - 4 | 176 | 74 | ||
| GenBank mRNA | AJ554300.1 | - | 2 - 3 | 81 | 74 | |
More Information
Set Membership: |
|

HCST
Genomic Map