Assay Details
| Gene Symbol: LPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448517.1 | XP_015304003.1 | 25 - 26 | 4378 | 66 | |
| XM_015448518.1 | XP_015304004.1 | 25 - 26 | 4378 | 66 | ||
| XM_015448519.1 | XP_015304005.1 | 23 - 24 | 4036 | 66 | ||
| XM_015448520.1 | XP_015304006.1 | 23 - 24 | 3834 | 66 | ||
More Information
Set Membership: |
|

LPA
Genomic Map