Assay Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587316.2 | XP_005587373.1 | 5 - 6 | 537 | 73 | |
| XM_015440341.1 | XP_015295827.1 | 5 - 6 | 504 | 73 | ||
| XM_015440342.1 | XP_015295828.1 | 5 - 6 | 456 | 73 | ||
More Information
Set Membership: |
|

RNF126
Genomic Map