Assay Details
| Gene Symbol: AHSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546616.2 | XP_005546673.1 | 6 - 7 | 980 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AHSG| Gene Symbol: AHSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546616.2 | XP_005546673.1 | 6 - 7 | 980 | 76 | |
Set Membership: |
|