Assay Details
| Gene Symbol: SLC16A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593967.2 | XP_005594024.1 | 1 - 2 | 617 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC16A2| Gene Symbol: SLC16A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593967.2 | XP_005594024.1 | 1 - 2 | 617 | 105 | |
Set Membership: |
|