Assay Details
| Gene Symbol: DCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581452.2 | XP_005581509.1 | 4 - 5 | 410 | 79 | |
| XM_015437060.1 | XP_015292546.1 | 4 - 5 | 410 | 79 | ||
More Information
Set Membership: |
|

DCTN3
Genomic Map