Assay Details
| Gene Symbol: ANKRD54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567334.2 | XP_005567391.1 | 4 - 5 | 901 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD54| Gene Symbol: ANKRD54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567334.2 | XP_005567391.1 | 4 - 5 | 901 | 89 | |
Set Membership: |
|