Assay Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587809.2 | XP_005587866.1 | 1 - 2 | 244 | 124 | |
| XM_015440386.1 | XP_015295872.1 | 1 - 2 | 251 | 124 | ||
More Information
Set Membership: |
|

SNAPC2
Genomic Map