Assay Details
| Gene Symbol: STOML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581411.2 | XP_005581468.1 | 5 - 6 | 535 | 81 | |
| XM_015437035.1 | XP_015292521.1 | 5 - 6 | 553 | 81 | ||
More Information
Set Membership: |
|

STOML2
Genomic Map