Assay Details
| Gene Symbol: GJB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585446.2 | XP_005585503.1 | 1 - 2 | 1038 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005585446.2 transcripts.
Set Membership: |
|

GJB6
Genomic Map