Assay Details
| Gene Symbol: HSD17B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284145.1 | NP_001271074.1 | 2 - 3 | 191 | 75 | |
| GenBank mRNA | DQ078723.1 | - | 2 - 3 | 191 | 75 | |
More Information
Set Membership: |
|

HSD17B10
Genomic Map