Assay Details
| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572557.2 | XP_005572614.1 | 1 - 2 | 713 | 100 | |
| XM_015431746.1 | XP_015287232.1 | 1 - 2 | 713 | 100 | ||
More Information
Set Membership: |
|

RILPL2
Genomic Map