Assay Details
| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553571.2 | XP_005553628.1 | 3 - 4 | 1501 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCF19| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553571.2 | XP_005553628.1 | 3 - 4 | 1501 | 82 | |
Set Membership: |
|