Assay Details
| Gene Symbol: SUCNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546129.2 | XP_005546186.1 | 2 - 3 | 165 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005546129.2 transcripts.
Set Membership: |
|

SUCNR1
Genomic Map