Assay Details
| Gene Symbol: TMEM95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582741.2 | XP_005582798.1 | 4 - 5 | 470 | 69 | |
| XM_015437573.1 | XP_015293059.1 | 3 - 4 | 413 | 69 | ||
More Information
Set Membership: |
|

TMEM95
Genomic Map