Assay Details
| Gene Symbol: TERF2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592589.2 | XP_005592646.1 | 1 - 2 | 838 | 96 | |
| XM_015442958.1 | XP_015298444.1 | 1 - 2 | 838 | 96 | ||
| GenBank mRNA | AB168345.1 | - | 1 - 2 | 777 | 96 | |
| AB169914.1 | - | 1 - 2 | 808 | 96 | ||
| AB170847.1 | - | 1 - 2 | 778 | 96 | ||
| AB171109.1 | - | 779 | 96 | |||
| AB171183.1 | - | 1 - 2 | 808 | 96 | ||
| AB171260.1 | - | 1 - 2 | 793 | 96 | ||
More Information
Set Membership: |
|

TERF2IP
Genomic Map