Assay Details
| Gene Symbol: PSMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283242.1 | NP_001270171.1 | 6 - 7 | 496 | 78 | |
| GenBank mRNA | AB168265.1 | - | 6 - 7 | 496 | 78 | |
| AB169649.1 | - | 6 - 7 | 513 | 78 | ||
More Information
Set Membership: |
|

PSMA4
Genomic Map