Assay Details
| Gene Symbol: HSDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592662.2 | XP_005592719.1 | 5 - 6 | 1070 | 73 | |
| XM_015442965.1 | XP_015298451.1 | 6 - 7 | 1113 | 73 | ||
More Information
Set Membership: |
|

HSDL1
Genomic Map