Assay Details
Gene Symbol: CNOT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575102.2 | XP_005575159.1 | 4 - 5 | 1214 | 93 | |
XM_015433278.1 | XP_015288764.1 | 3 - 4 | 1049 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
