Assay Details
Gene Symbol: FAM162A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547992.2 | XP_005548049.1 | 4 - 5 | 503 | 67 | |
XM_005547993.2 | XP_005548050.1 | 4 - 5 | 403 | 67 | ||
XM_015446356.1 | XP_015301842.1 | 5 - 6 | 585 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
