Assay Details
| Gene Symbol: DDIT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283340.1 | NP_001270269.1 | 2 - 3 | 405 | 68 | |
| XM_015455864.1 | XP_015311350.1 | 1 - 2 | 499 | 68 | ||
| GenBank mRNA | AB171014.1 | - | 2 - 3 | 405 | 68 | |
More Information
Set Membership: |
|

DDIT4
Genomic Map