Assay Details
Gene Symbol: USP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592681.2 | XP_005592738.1 | 9 - 10 | 1782 | 105 | |
XM_005592682.2 | XP_005592739.1 | 9 - 10 | 1781 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
