Assay Details
| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548649.2 | XP_005548706.1 | 3 - 4 | 333 | 72 | |
| XM_005548653.2 | XP_005548710.1 | 3 - 4 | 657 | 72 | ||
More Information
Set Membership: |
|

FAM3B
Genomic Map