Assay Details
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545863.2 | XP_005545920.1 | 4 - 5 | 835 | 56 | |
| XM_015445129.1 | XP_015300615.1 | 3 - 4 | 502 | 56 | ||
More Information
Set Membership: |
|

CLDN18
Genomic Map